Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ptprc, Mouse, Clone: ANK74, Abnova™

Catalog No. 89112201 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-201 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-201 Supplier Abnova Corporation Supplier No. MAB5278
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Ptprc.

Specifications

Antigen Ptprc
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ANK74
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Ptprc.
Dilution The optimal working dilution should be determined by the end user.
Formulation In culture supernatant (0.05% sodium azide)
Gene Ptprc
Gene Alias CD45/L-CA/Lca/RT7/T200
Gene Symbols Ptprc
Host Species Mouse
Immunogen Native purified Ptprc from IL-2-activated cultured Wag rats NK cells.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24699
Test Specificity This is a rat strain-independent antibody to CD45. This monoclonal antibody recognizes a common epitope of rat CD45, present on all rat leucocytes.
Target Species Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.