Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ptprc, Mouse, Clone: HIS24, Abnova™

Catalog No. 89109183 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-183 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-183 Supplier Abnova Corporation Supplier No. MAB10968

Mouse monoclonal antibody raised against Ptprc.

Specifications

Antigen Ptprc
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone HIS24
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Ptprc.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Ptprc
Gene Alias CD45/L-CA/Lca/RT7/T200
Gene Symbols Ptprc
Host Species Mouse
Immunogen Rat Ptprc.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24699
Test Specificity The monoclonal antibody reacts with an isoform of Ptprc expressed by rat B cells.
Target Species Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.