Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRR, Mouse, Clone: 6A6, Abnova™

Catalog No. 89112340 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-340 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-340 Supplier Abnova Corporation Supplier No. MAB5645
Add to Cart
Add to Cart
Request a Quote

Mouse monoclonal antibody raised against recombinant PTPRR.

Specifications

Antigen PTPRR
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6A6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant PTPRR.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM ammonium bicarbonate buffer (2mg BSA)
Gene Ptprr
Gene Alias Gmcp1/PTP-SL/PTPBR7/mPTP213
Gene Symbols Ptprr
Host Species Mouse
Immunogen Recombinant GST fusion protein corresponding to mouse PTPRR.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19279
Test Specificity Protein tyrosine phosphatases (PTPs) are the enzymes that are instrumental in determining the spatial and temporal balance between the tyrosine phosphorylated and non-phosphorylated targets, and thus coordinately regulate these cellular responses to extracellular cues. The Ptprr gene gives rise to 4 different neuronal phosphatases which differ in length of their N-terminal part and subcellular localization. PTPBR7 (72 KDa) is receptor-like, PTP-SL (60 KDa) is membrane associated and PTPPBSgamma (42 and 37 KDa) is a cytosolic phoaphatase. This antibody is directed to the common part of these proteins.
Target Species Mouse
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.