Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PXN, Mouse, Clone: M107, Abnova™

Catalog No. 89110382 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-382 Supplier Abnova Corporation Supplier No. MAB1366
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant PXN.

Specifications

Antigen PXN
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M107
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant PXN.
Dilution ELISA (1:2000) Immunocytochemistry (1:100) Immunoprecipitation (1-5 ul) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Gene PXN
Gene Alias FLJ16691
Gene Symbols PXN
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of human PXN.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5829
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.