Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-PXR, Clone: 1F10.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABC287
Description
Specifically detects PXR clone: 1F10.1 in Human samples, and it is validated for Western Blotting
Nuclear receptor subfamily 1 group I member 2 (UniProt O75469; also known as Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, Pregnane X nuclear receptor, Steroid and xenobiotic receptor, SXR) is encoded by the NR1I2 (also known as BXR, PAR, PRR, PXR, SAR, SXR, ONR1) gene (Gene ID 8856) in human. Pregnane X receptor (PXR) is a nuclear receptor that displays similarity to PPAR and RXR. PXR is activated by a large number of endogenous and exogenous chemicals including steroids and antibiotics. One of the key roles of PXR is the induction of CYP3A4, a P450 enzyme involved in drug metabolism.
Specifications
PXR | |
Monoclonal | |
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
NR1I2; PXR | |
KLH-conjugated linear peptide corresponding to human PXR. | |
100 μg | |
Apoptosis, Cancer | |
NP_003880 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG2b κ |
Western Blot | |
1F10.1 | |
O75469 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction