Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qa2, Mouse, PE, Clone: 695H1-9-9, Abnova™

Catalog No. 89112477 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-477 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-477 Supplier Abnova Corporation Supplier No. MAB5813
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Qa2.

Specifications

Antigen Qa2
Applications Flow Cytometry
Classification Monoclonal
Clone 695H1-9-9
Conjugate PE
Description Mouse monoclonal antibody raised against native Qa2.
Dilution Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Qa2
Gene Alias H2-Qa2/Ped/Qa-2
Gene Symbols Qa2
Host Species Mouse
Immunogen Native purified from C3H.SW mouse skin graft and splenocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 111458
Test Specificity Qa-2 alloantigen.
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.