Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB2A, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89109346 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-346 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-346 Supplier Abnova Corporation Supplier No. MAB11163
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant protein of RAB2A.

Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rabs are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking. The mammalian RAB proteins show striking similarities to the S. cerevisiae YPT1 and SEC4 proteins, Ras-related GTP-binding proteins involved in the regulation of secretion.[supplied by OMIM

Specifications

Antigen RAB2A
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant protein of RAB2A.
Dilution ELISA (1:5000-1:20000) Western Blot (1:100-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (1% BSA, 50% glycerol, 0.02% sodium azide)
Gene RAB2A
Gene Alias RAB2
Gene Symbols RAB2A
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminus of human RAB2A.
Purification Method Protein G purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5862
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.