Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB3D, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106101 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-101 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-101 Supplier Abnova Corporation Supplier No. H00009545A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant RAB3D.

Sequence: MASAGDTQAGPRDAADQNFDYMFKLLLIGNSSVGKTSFLFRYADDSFTPAFVSTVGIDFKVKTVYRHDKRIKLQIWDTAGQERYRTITTAYYRGAMGFLLMYDIANQESFAAVQDWATQIKTYSWDNAQVILVGNKCDLEDERVVPAEDGRRLADDLGFEFFEASAKENINVKQVFERLVDVICEKMNESLEPSSSSGSNGKGPAVGDAPAPQPSSCSC

Specifications

Antigen RAB3D
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant RAB3D.
Formulation 50% glycerol
Gene RAB3D
Gene Accession No. BC016471
Gene Alias D2-2/GOV/RAB16/RAD3D
Gene Symbols RAB3D
Host Species Mouse
Immunogen RAB3D (AAH16471, 1 a.a. ∽ 219 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9545
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.