Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Human IgG Secondary Antibody (MR12), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250261AF750
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Immunohistochemistry.Specifications
Rabbit IgG | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
Immunoglobulin purified from rabbit serum | |
0.1 mL | |
Human, Rabbit | |
IgG1 |
Immunohistochemistry | |
MR12 | |
Immunohistochemistry | |
Mouse | |
Protein A purified | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction