Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit Light Chain, Mouse, Clone: SB62a, Abnova™

Catalog No. 89135421 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-421 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-421 Supplier Abnova Corporation Supplier No. MAB3268
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Rabbit Light Chain.

Specifications

Antigen Rabbit Light Chain
Applications ELISA, Western Blot
Classification Monoclonal
Clone SB62a
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Rabbit Light Chain.
Dilution ELISA (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Host Species Mouse
Immunogen Rabbit Light Chain.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Rabbit Light Chain.
Target Species Rabbit
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.