Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABIF, Mouse, Clone: 2G3, Abnova™

Catalog No. 89105270 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-270 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-270 Supplier Abnova Corporation Supplier No. H00005877M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant RABIF.

The Sec4/Rab-related small GTP-binding proteins are involved in the regulation of intracellular vesicular transport. Mss4 stimulates GTP-GDP exchange in Sec4 and Rab and binds to a subset of genetically related Rab proteins.[supplied by OMIM

Sequence: MEPAEQPSELVSAEGRNRKAVLCQRCGSRVLQPGTALFSRRQLFLPSMRKKPALSDGSNPDGDLLQEHWLVEDMFIFENVGFTKDVGNIKFLVCADCEIGPIGWHCLDDKNSFYVALERVSHE

Specifications

Antigen RABIF
Applications ELISA
Classification Monoclonal
Clone 2G3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant RABIF.
Formulation ascites with no preservative
Gene RABIF
Gene Accession No. BC018488
Gene Alias MSS4/RASGFR3/RASGRF3
Gene Symbols RABIF
Host Species Mouse
Immunogen RABIF (AAH18488,1 a.a. ∽ 123 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5877
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.