Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD21, Mouse, Clone: CM110-2C10, Abnova™

Catalog No. 89110915 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-915 Supplier Abnova Corporation Supplier No. MAB2300
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant RAD21.

The protein encoded by this gene is highly similar to the gene product of Schizosaccharomyces pombe rad21, a gene involved in the repair of DNA double-strand breaks, as well as in chromatid cohesion during mitosis. This protein is a nuclear phospho-protein, which becomes hyperphosphorylated in cell cycle M phase. The highly regulated association of this protein with mitotic chromatin specifically at the centromere region suggests its role in sister chromatid cohesion in mitotic cells. [provided by RefSeq

Specifications

Antigen RAD21
Applications Western Blot
Classification Monoclonal
Clone CM110-2C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant RAD21.
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene RAD21
Gene Alias FLJ25655/FLJ40596/HR21/HRAD21/KIAA0078/MCD1/NXP1/SCC1/hHR21
Gene Symbols RAD21
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 544-631 of human RAD21.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5885
Test Specificity This antibody is specific to both the nuclear hRad21 protein and the cleaved cytoplasmic hRad21 protein.
Target Species Human, Mouse
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.