Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Rad51, Clone: 3C10,

Catalog No. 05530IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-530-IMI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-530-IMI Supplier MilliporeSigma™ Supplier No. 05530I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Rad51 clone: 3C10 in Human samples, and it is validated for Immunoprecipitation, Western Blotting

DNA repair protein RAD51 homolog 1 (UniProt Q06609; also known as BRCA1/BRCA2-containing complex subunit 5, Homolog of E. coli RecA, hRAD51, HsRAD51, RAD51 homolog A, RecA-like protein, Recombination protein A) is encoded by the RAD51 (also known as HRAD51, HsT16930, BRCC5, MRMV2, RAD51A, RECA) gene (Gene ID 5888) in human. RAD51 is a member of the RecA family recombinases. It interacts with BRCA1 and BRCA2 to activate homologous recombination. RAD51 exhibits DNA-dependent ATPase activity and participates in double-strand break (DSB) repair by unwinding DNA to form a helical nucleoprotein filament. It is highly expressed in the testis and thymus and lower expression levels are found in the small intestine, placenta, colon, pancreas, and ovaries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rad51
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3C10
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q06609
Gene Symbols RAD51; RAD51A; RECA
Host Species Mouse
Immunogen Recombinant protein corresponding to human Rad51.
Purification Method Protein G Purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_002866
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.