Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD51L1, Mouse, Clone: 1 H3/13, Abnova™

Catalog No. 89110988 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-988 Supplier Abnova Corporation Supplier No. MAB2396
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant RAD51L1.

The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are evolutionarily conserved proteins essential for DNA repair by homologous recombination. This protein has been shown to form a stable heterodimer with the family member RAD51C, which further interacts with the other family members, such as RAD51, XRCC2, and XRCC3. Overexpression of this gene was found to cause cell cycle G1 delay and cell apoptosis, which suggested a role of this protein in sensing DNA damage. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq

Specifications

Antigen RAD51L1
Applications Western Blot
Classification Monoclonal
Clone 1 H3/13
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant RAD51L1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene RAD51L1
Gene Alias MGC34245/R51H2/RAD51B/REC2/hREC2
Gene Symbols RAD51L1
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to human RAD51L1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5890
Test Specificity This antibody is specific to Rad51B.
Target Species Human, Primate
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.