Learn More
RAD51L3, Mouse, Clone: 5B3/6, Abnova™
Mouse monoclonal antibody raised against recombinant RAD51L3.
Supplier: Abnova Corporation MAB2395
Description
The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are highly similar to bacterial RecA and Saccharomyces cerevisiae Rad51, which are known to be involved in the homologous recombination and repair of DNA. This protein forms a complex with several other members of the RAD51 family, including RAD51L1, RAD51L2, and XRCC2. The protein complex formed with this protein has been shown to catalyze homologous pairing between single- and double-stranded DNA, and is thought to play a role in the early stage of recombinational repair of DNA. Several alternatively spliced transcript variants of this gene have been described, but the biological validity of some of them has not been determined. [provided by RefSeq
Specifications
RAD51L3 | |
Monoclonal | |
Unconjugated | |
Immunocytochemistry (1:100) Immunofluorescence (1:500) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
RAD51L3 | |
RAD51L3 | |
Recombinant His fusion protein corresponding to human RAD51L3. | |
RUO | |
5892 | |
Human | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
5B3/6 | |
Mouse monoclonal antibody raised against recombinant RAD51L3. | |
In PBS | |
HsTRAD/R51H3/RAD51D/Trad | |
Mouse | |
100 μL | |
Primary | |
This antibody is specific to Rad51D. | |
Store at 4°C. Do not freeze. | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.