Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAF1, Mouse, Clone: M208, Abnova™

Catalog No. 89113035 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-035 Supplier Abnova Corporation Supplier No. MAB6771
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of RAF1.

This gene is the cellular homolog of viral raf gene (v-raf). The encoded protein is a MAP kinase kinase kinase (MAP3K), which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated, the cellular RAF1 protein can phosphorylate to activate the dual specificity protein kinases MEK1 and MEK2, which in turn phosphorylate to activate the serine/threonine specific protein kinases, ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration. Mutations in this gene are associated with Noonan syndrome 5 and LEOPARD syndrome 2. [provided by RefSeq

Specifications

Antigen RAF1
Applications ELISA, Western Blot
Classification Monoclonal
Clone M208
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of RAF1.
Dilution ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Gene RAF1
Gene Alias CRAF/NS5/Raf-1/c-Raf
Gene Symbols RAF1
Host Species Mouse
Immunogen A synthetic peptide corresponding to N-terminus of human RAF1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5894
Test Specificity This C-Raf sequence has high homology to rat and mouse C-Raf.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.