Learn More
CD32 Mouse anti-Rat, BUV395, Clone: D34-485, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 740263
Description
The D34-485 monoclonal antibody specifically recognizes CD32, the Fc γII receptor. Rat CD32 is expressed on B lymphocytes, myeloid cells, and some lymphocytes in the thymic medulla. D34-485 antibody blocks binding of aggregated immunoglobulins to the Fc γII receptors in vitro.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348nm and an Em Max at 395nm, BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

Specifications
CD32 | |
Monoclonal | |
0.2mg/mL | |
Fcgr2b; FcRII; Fc-gamma RII; FcγRII; FcγII receptor | |
Mouse | |
Affinity Purified | |
RUO | |
Rat | |
IgG1 κ |
Flow Cytometry | |
D34-485 | |
Brilliant Ultraviolet 395 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant Rat CD32 Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only