Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Tumors, epithelial origin, Mouse, Clone: CC52, Abnova™

Catalog No. 89135917 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-917 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-917 Supplier Abnova Corporation Supplier No. MAB5199
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Rat Tumors, epithelial origin.

Specifications

Antigen Rat Tumors, epithelial origin
Applications Western Blot
Classification Monoclonal
Clone CC52
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Rat Tumors, epithelial origin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.05% sodium azide
Host Species Mouse
Immunogen Native purified from rat colon adenocarcinoma cells (CC531).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CC52 is a rat strain-independent marker for tumor cells of epithelial origin, such as colon, breast, or lung cancer, etc. When injected in colon tumor-bearing rats, CC52 localized to tumor cells (Hagenaars et al., 2001). CC52 can be used for immunohistochemical staining of tumor cells in frozen tissue sections as well as formalin-fixed/paraffin-embedded tissue. CC52 is also suitable for flow cytometry, Western blotting, immunoprecipitation, and ELISA. Hybridomas were selected for producing antibodies reacting with the CC531 cell line and with minimal cross reactivity to normal tissues (Hagenaars et al., 2001). CC52 belongs to a first generation of such antibodies and is suitable to detect tumor cells by immunohistochemistry. The monoclonal antibody binds to a dimer of two proteins, 120 kD, 130 kD, expressed by rat tumor cells of epithelial origin. CC52 has been used in many studies in a rat model of colorectal cancer (Hagenaars et al., 2000). In immunohistochemical stainings, there is
Target Species Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.