Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB1, Mouse, Clone: 1D1, Abnova™

Catalog No. 89110864 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-864 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-864 Supplier Abnova Corporation Supplier No. MAB2168
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant RB1.

The protein encoded by this gene is a negative regulator of the cell cycle and was the first tumor suppressor gene found. The encoded protein also stabilizes constitutive heterochromatin to maintain the overall chromatin structure. The active, hypophosphorylated form of the protein binds transcription factor E2F1. Defects in this gene are a cause of childhood cancer retinoblastoma (RB), bladder cancer, and osteogenic sarcoma. [provided by RefSeq

Specifications

Antigen RB1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1D1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant RB1.
Dilution ELISA Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene RB1
Gene Alias OSRC/RB/p105-Rb/pRb/pp110
Gene Symbols RB1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 571-590 of RB1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5925
Test Specificity This antibody recognizes an epitope within aa 571-590 of RB1.
Target Species Human
Content And Storage Store at -20°C to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.