Learn More
RB1, Mouse, Clone: 3C8, Abnova™
Mouse monoclonal antibody raised against partial recombinant RB1.
Supplier: Abnova Corporation MAB2169
Description
The protein encoded by this gene is a negative regulator of the cell cycle and was the first tumor suppressor gene found. The encoded protein also stabilizes constitutive heterochromatin to maintain the overall chromatin structure. The active, hypophosphorylated form of the protein binds transcription factor E2F1. Defects in this gene are a cause of childhood cancer retinoblastoma (RB), bladder cancer, and osteogenic sarcoma. [provided by RefSeq
Specifications
RB1 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
RB1 | |
RB1 | |
Recombinant protein corresponding to amino acids 886-905 of RB1. | |
RUO | |
5925 | |
Human | |
Liquid |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
3C8 | |
Mouse monoclonal antibody raised against partial recombinant RB1. | |
In PBS, pH 7.4 | |
OSRC/RB/p105-Rb/pRb/pp110 | |
Mouse | |
100 μg | |
Primary | |
This antibody recognizes an epitope within aa 886-905 of RB1. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.