Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP6, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105303 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-303 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-303 Supplier Abnova Corporation Supplier No. H00005930A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant RBBP6.

The retinoblastoma tumor suppressor (pRB) protein binds with many other proteins. In various human cancers, pRB suppresses cellular proliferation and is inactivated. Cell cycle-dependent phosphorylation regulates the activity of pRB. This gene encodes a protein which binds to underphosphorylated but not phosphorylated pRB. Multiple alternatively spliced transcript variants that encode different isoforms have been found for this gene. [provided by RefSeq

Sequence: SSGNKLLYILNPPETQVEKEQITGQIDKSTVKPKPQLSHSSRLSSDLTRETDEAAFEPDYNESDSESNVSVKEEESSGNISKDLKDKIVEKAKESLDTAAVVQVGISRNQ

Specifications

Antigen RBBP6
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant RBBP6.
Formulation 50% glycerol
Gene RBBP6
Gene Accession No. NM_006910
Gene Alias DKFZp686P0638/DKFZp761B2423/MY038/P2P-R/PACT/RBQ-1/SNAMA
Gene Symbols RBBP6
Host Species Mouse
Immunogen RBBP6 (NP_008841, 1582 a.a. ∽ 1691 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5930
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.