Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCN2, Mouse, Clone: 2B12, Abnova™

Catalog No. 89105308 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-308 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-308 Supplier Abnova Corporation Supplier No. H00005955M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant RCN2.

The protein encoded by this gene is a calcium-binding protein located in the lumen of the ER. The protein contains six conserved regions with similarity to a high affinity Ca(+2)-binding motif, the EF-hand. This gene maps to the same region as type 4 Bardet-Biedl syndrome, suggesting a possible causative role for this gene in the disorder. [provided by RefSeq]

Sequence: KAEELHYPLGERRSDYDREALLGVQEDVDEYVKLGHEEQQKRLQAIIKKIDLDSDGFLTESELSSWIQMSFKHYAMQEAKQQFVEYDKNSDDTVTWDEYNIQMYDRVIDFDENTALDDAEEESFRKLHLKDKKRFEKANQDSGPGLSLEEFIAFEHPEEVDYMTEFVIQEALEEHDKNGDGFVSLEEFLGDYRWDPTANEDPEWILVEKDRFVNDYDKDNDGRLDPQELLPWVVPNNQGIAQEEALHLIDEMDLNGDKKLSEEEILENPDLFLTSEATDYGRQLHDDYFYHDEL

Specifications

Antigen RCN2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant RCN2.
Formulation ascites with no preservative
Gene RCN2
Gene Accession No. BC004892
Gene Alias E6BP/ERC-55/ERC55/TCBP49
Gene Symbols RCN2
Host Species Mouse
Immunogen RCN2 (AAH04892.1,24 a.a. ∽ 317 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5955
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.