Learn More
RCV1, Mouse, Clone: 1D3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RCV1.
Supplier: Abnova Corporation H00005957M03A
Description
This gene encodes a member of the recoverin family of neuronal calcium sensors. The encoded protein contains three calcium-binding EF-hand domains and may prolong the termination of the phototransduction cascade in the retina by blocking the phosphorylation of photo-activated rhodopsin. Recoverin may be the antigen responsible for cancer-associated retinopathy. [provided by RefSeq
Sequence: KLEWAFSLYDVDGNGTISKNEVLEIVMAIFKMITPEDVKLLPDDENTPEKRAEKIWKYFGKNDDDKLTEKEFIEGTLANKEILRLIQFEPQKVKEKMKNSpecifications
RCV1 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_002903.1 | |
RCVRN | |
RCV1 (NP_002894.1,101 a.a. ∽ 199 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
5957 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Immunoprecipitation, Western Blot | |
1D3 | |
Mouse monoclonal antibody raised against a partial recombinant RCV1. | |
RCVRN | |
RCV1 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.