Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDX, Mouse, Clone: 1E12, Abnova™

Catalog No. 89105318 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-318 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-318 Supplier Abnova Corporation Supplier No. H00005962M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant RDX.

Radixin is a cytoskeletal protein that may be important in linking actin to the plasma membrane. It is highly similar in sequence to both ezrin and moesin. The radixin gene has been localized by fluorescence in situ hybridization to 11q23. A truncated version representing a pseudogene (RDXP2) was assigned to Xp21.3. Another pseudogene that seemed to lack introns (RDXP1) was mapped to 11p by Southern and PCR analyses. [provided by RefSeq

Sequence: MPKPINVRVTTMDAELEFAIQPNTTGKQLFDQVVKTVGLREVWFFGLQYVDSKGYSTWLKLNKKVTQQDVKKENPLQFKFRAKFFPEDVSEELIQEITQRLFFLQVKEAILNDEIYCPPETAVLLASYAVQAKYGDYNKEIHKPGYLANDRLLPQRVLEQHKLTKEQWEERIQNWHEEHRGMLREDSMMEYLKIAQDLEMYGVNYFEIKNKKGTELWLGVDALGLNIYEHDDKLTPKIGFPWSEIRNISFNDKKFVIKPIDKKAPDFVFYAPRLRINKRILALCMGNHELYMRRRKPDTIEVQQMKAQAREEKHQKQLERAQLENEKEKREIAEKEKERIEREKEELMERLKQIEEQTIKAQKELEEQTRKALELDQERKRAKEEAERLEKERRAAEEAKSAIAKQAADQMKNQEQLAAELAEFTAKIALLEEAKKKKEEEATEWQHKAFAAQEDLEKTKEELKTVMSAPPPPPPPPVIPPTENEHDEHDENNAEASAELSNEGVMNHRSEEERVTETQKNERVKKQLQALSSELAQARDETKKTQNDVLHAENVKAGRDKYKTLRQIRQGNTKQRIDEFEAM

Specifications

Antigen RDX
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant RDX.
Formulation ascites with no preservative
Gene RDX
Gene Accession No. BC047109
Gene Alias DFNB24
Gene Symbols RDX
Host Species Mouse
Immunogen RDX (AAH47109,1 a.a. ∽ 583 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5962
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.