Learn More
REG1A, Mouse, Clone: 1A4, Abnova™
Mouse monoclonal antibody raised against recombinant REG1A.
Supplier: Abnova Corporation MAB10312
Description
This gene is a type I subclass member of the Reg gene family. The Reg gene family is a multigene family grouped into four subclasses, types I, II, III and IV, based on the primary structures of the encoded proteins. This gene encodes a protein that is secreted by the exocrine pancreas. It is associated with islet cell regeneration and diabetogenesis and may be involved in pancreatic lithogenesis. Reg family members REG1B, REGL, PAP and this gene are tandemly clustered on chromosome 2p12 and may have arisen from the same ancestral gene by gene duplication. [provided by RefSeq
Specifications
REG1A | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
REG1A | |
REG1A | |
Recombinant protein corresponding to human REG1A. | |
RUO | |
5967 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Immunofluorescence, Immunohistochemistry | |
1A4 | |
Mouse monoclonal antibody raised against recombinant REG1A. | |
In ascites (0.03% sodium azide) | |
ICRF/MGC12447/P19/PSP/PSPS/PSPS1/PTP/REG | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.