Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory syncytial virus Nucleoprotein, Mouse, FITC, Clone: 671, Abnova™

Catalog No. 89135998 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-998 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-998 Supplier Abnova Corporation Supplier No. MAB5510
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Respiratory syncytial virus Nucleoprotein.

Specifications

Antigen Respiratory syncytial virus Nucleoprotein
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 671
Conjugate FITC
Description Mouse monoclonal antibody raised against Respiratory syncytial virus Nucleoprotein.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (10mg/mL BSA, 0.09% sodium azide)
Host Species Mouse
Immunogen Native from respiratory syncytial virus viral lysate (Long strain).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for the nucleoprotein of RSV.
Target Species Respiratory Syncytial virus (RSV)
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.