Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ret Oncoprotein, Mouse, Clone: 3F8, Abnova™

Catalog No. 89136753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-753 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-753 Supplier Abnova Corporation Supplier No. MAB9697

Mouse monoclonal antibody raised against partial recombinant ret Oncoprotein.

Specifications

Antigen ret Oncoprotein
Applications Immunohistochemistry
Classification Monoclonal
Clone 3F8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ret Oncoprotein.
Dilution Immunohistochemistry (1:20-1:40) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from tissue culture supernatant (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to the extreme C-terminal cytoplasmic region of human ret Oncoprotein.
Quantity 3 x 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with sterile distilled water store at 4°C for at least two months. For long term storage store at -20°C or below. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.