Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RETN, Mouse, Clone: 4B11, Abnova™

Catalog No. 89107144 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-144 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-144 Supplier Abnova Corporation Supplier No. H00056729M24A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant RETN.

This gene belongs to the family defined by the mouse resistin-like genes. The characteristic feature of this family is the C-terminal stretch of 10 cys residues with identical spacing. The mouse homolog of this protein is secreted by adipocytes, and may be the hormone potentially linking obesity to type II diabetes. [provided by RefSeq

Sequence: TLCSMEEAINERIQEVAGSLIFRAISSIGLECQSVTSRGDLATCPRGFAVTGCTCGSACGSWDVRAETTCHCQCAGMDWTGARCCRV*

Specifications

Antigen RETN
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4B11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant RETN.
Formulation ascites with no preservative
Gene RETN
Gene Accession No. NM_020415
Gene Alias ADSF/FIZZ3/MGC126603/MGC126609/RETN1/RSTN/XCP1
Gene Symbols RETN
Host Species Mouse
Immunogen RETN (NP_065148,20 a.a. ∽ 106 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56729
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.