Learn More
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antigen | RFP |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 1610 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:500-1:1000 |
| Formulation | PBS with No Preservative |
| Gene Alias | DSRED, GFP-like chromoprotein, Red Fluorescent Protein, RFP |
| Host Species | Mouse |
| Immunogen | Recombinant protein conjugated synthetic peptide encompassing a sequence within the center region of RFP. The exact sequence is proprietary. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
