Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHO, Mouse, Clone: 4D2, Abnova™

Catalog No. 89113494 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-494 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-113-494 Supplier Abnova Corporation Supplier No. MAB8558
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against full-length native RHO.

Specifications

Antigen RHO
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4D2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length native RHO.
Dilution Western Blot (1:1000) Immunoprecipitation(1-2 ug/100-500 ug protein) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene RHO
Gene Symbols RHO
Host Species Mouse
Immunogen Native purified bovine RHO.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 509933
Test Specificity This antibody is specific to N-terminus of RHO.
Target Species Bovine, Mammalia
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.