Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIN3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107347 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-347 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-347 Supplier Abnova Corporation Supplier No. H00079890A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant RIN3.

RIN3 is a member of the RIN family of Ras interaction-interference proteins, which are binding partners to the RAB5 small GTPases (see RAB5A; MIM 179512) (Kajiho et al., 2003 [PubMed 12972505]).[supplied by OMIM

Sequence: GPPDHPNQPPMMTCERLPCPTAGLGPLREEAMKPGAASSPLQQVPAPPLPAKKNLPTAPPRRRVSERVSLEDQSPGMAAEGDQLSLPPQGTSDGPEDTP

Specifications

Antigen RIN3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant RIN3.
Formulation 50% glycerol
Gene RIN3
Gene Accession No. NM_024832
Gene Alias DKFZp762H1613/FLJ11700/FLJ22439
Gene Symbols RIN3
Host Species Mouse
Immunogen RIN3 (NP_079108, 328 a.a. ∽ 426 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79890
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.