Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RING1, Mouse, Clone: 2B3, Abnova™

Catalog No. 89105335 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-335 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-335 Supplier Abnova Corporation Supplier No. H00006015M03J
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant RING1.

This gene belongs to the RING finger family, members of which encode proteins characterized by a RING domain, a zinc-binding motif related to the zinc finger domain. The gene product can bind DNA and can act as a transcriptional repressor. It is associated with the multimeric polycomb group protein complex. The gene product interacts with the polycomb group proteins BMI1, EDR1, and CBX4, and colocalizes with these proteins in large nuclear domains. It interacts with the CBX4 protein via its glycine-rich C-terminal domain. The gene maps to the HLA class II region, where it is contiguous with the RING finger genes FABGL and HKE4. [provided by RefSeq

Sequence: NKECPTCRKKLVSKRSLRPDPNFDALISKIYPSREEYEAHQDRVLIRLSRLHNQQALSSSIEEGLRMQAMHRAQRVRRPIPGSDQTTTMS

Specifications

Antigen RING1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant RING1.
Formulation PBS with no preservative; pH 7.4
Gene RING1
Gene Accession No. NM_002931
Gene Alias RING1A/RNF1
Gene Symbols RING1
Host Species Mouse
Immunogen RING1 (NP_002922,81 a.a. ∽ 170 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6015
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.