Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK3, Mouse, Clone: Rippy-3, Abnova™

Catalog No. 89111567 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-567 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-567 Supplier Abnova Corporation Supplier No. MAB3728
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant RIPK3.

The product of this gene is a member of the receptor-interacting protein (RIP) family of serine/threonine protein kinases, and contains a C-terminal domain unique from other RIP family members. The encoded protein is predominantly localized to the cytoplasm, and can undergo nucleocytoplasmic shuttling dependent on novel nuclear localization and export signals. It is a component of the tumor necrosis factor (TNF) receptor-I signaling complex, and can induce apoptosis and weakly activate the NF-kappaB transcription factor. [provided by RefSeq

Specifications

Antigen RIPK3
Applications Western Blot
Classification Monoclonal
Clone Rippy-3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant RIPK3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene RIPK3
Gene Alias RIP3
Gene Symbols RIPK3
Host Species Mouse
Immunogen Recombinant protein corresponding to human RIPK3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11035
Test Specificity Recognizes human RIP3.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.