Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIT2, Mouse, Clone: 27G2, Abnova™

Catalog No. 89110757 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-757 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-757 Supplier Abnova Corporation Supplier No. MAB2012
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant RIT2.

RIN belongs to the RAS (HRAS; MIM 190020) superfamily of small GTPases (Shao et al., 1999 [PubMed 10545207]).[supplied by OMIM

Specifications

Antigen RIT2
Applications Western Blot
Classification Monoclonal
Clone 27G2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant RIT2.
Dilution Western blot (0.5 to 1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM sodium phosphate buffer, 150mM NaCl, pH 7.5 (50% glycerol, 3mM sodium azide)
Gene RIT2
Gene Alias RIBA/RIN/ROC2
Gene Symbols RIT2
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human RIT2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6014
Test Specificity This antibody detects recombinant Rin protein. Does not cross-react with Rit protein.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.