Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA Polymerase II, Mouse, Clone: 8WG16, Abnova™

Catalog No. 89136453 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-453 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-136-453 Supplier Abnova Corporation Supplier No. MAB8286
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against native RNA Polymerase II.

Specifications

Antigen RNA Polymerase II
Classification Monoclonal
Clone 8WG16
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native RNA Polymerase II.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Mouse
Immunogen Native purified RNA Polymerase II .
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with the highly conserved heptapeptide repeat of the largest subunit of eukaryotic RNA polymerase II. It inhibits promoterdirected transcription, but it does not inhibit elongation in the nonspecific transcription assay.
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.