Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNASEH2A, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106346 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-346 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-346 Supplier Abnova Corporation Supplier No. H00010535A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant RNASEH2A.

The protein encoded by this gene is a component of the heterotrimeric type II ribonuclease H enzyme (RNAseH2). RNAseH2 is the major source of ribonuclease H activity in mammalian cells and endonucleolytically cleaves ribonucleotides. It is predicted to remove Okazaki fragment RNA primers during lagging strand DNA synthesis and to excise single ribonucleotides from DNA-DNA duplexes. Mutations in this gene cause Aicardi-Goutieres Syndrome (AGS), a an autosomal recessive neurological disorder characterized by progressive microcephaly and psychomotor retardation, intracranial calcifications, elevated levels of interferon-alpha and white blood cells in the cerebrospinal fluid

Sequence: MDLSELERDNTGRCRLSSPVPAVCRKEPCVLGVDEAGRGPVLGPMVYAICYCPLPRLADLEALKVADSKTLLESERERLFAKMEDTDFVGWALDVLSPNLISTSMLGRVKYNLNSLSHDTATGLIQYALDQGVNVTQVFVDTVGMPETYQARLQQSFPGIEVTVKAKADALYPVVSAASICAKVARDQAVKKWQFVEKLQDLDTDYGSGYPNDPKTKAWLKEHVEPVFGFPQFVRFSWRTAQTILEKEAEDVIWEDSASENQEGLRKITSYFLNEGSQARPRSSHRYFLERGLESATSL

Specifications

Antigen RNASEH2A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant RNASEH2A.
Formulation 50% glycerol
Gene RNASEH2A
Gene Accession No. BC011748
Gene Alias AGS4/JUNB/RNASEHI/RNHIA/RNHL
Gene Symbols RNASEH2A
Host Species Mouse
Immunogen RNASEH2A (AAH11748, 1 a.a. ∽ 299 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10535
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.