Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNASEL, Mouse, Clone: 2E9, Abnova™

Catalog No. 89110975 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-975 Supplier Abnova Corporation Supplier No. MAB2378
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant RNASEL.

This gene encodes a component of the interferon-regulated 2-5A system that functions in the antiviral and antiproliferative roles of interferons. Mutations in this gene have been associated with predisposition to prostate cancer and this gene is a candidate for the hereditary prostate cancer 1 (HPC1) allele. [provided by RefSeq

Specifications

Antigen RNASEL
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant RNASEL.
Dilution Western Blot (1:1000-1:4000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene RNASEL
Gene Alias DKFZp781D08126/MGC104972/MGC133329/PRCA1/RNS4
Gene Symbols RNASEL
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human RNASEL.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6041
Test Specificity This antibody is specific to human RNaseL.
Target Species Human, Primate
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.