Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF122, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107343 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-343 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-343 Supplier Abnova Corporation Supplier No. H00079845A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant RNF122.

The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. [provided by RefSeq

Sequence: SKLRNQAQSERYGYKEVVLKGDAKKLQLYGQTCAVCLEDFKGKDELGVLPCQHAFHRKCLVKWLEVRCVCPMCNKPIASPSEATQNIGILLDELV

Specifications

Antigen RNF122
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant RNF122.
Formulation 50% glycerol
Gene RNF122
Gene Accession No. NM_024787
Gene Alias FLJ12526/MGC126622
Gene Symbols RNF122
Host Species Mouse
Immunogen RNF122 (NP_079063, 61 a.a. ∽ 155 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79845
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.