Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF13, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106514 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-514 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-514 Supplier Abnova Corporation Supplier No. H00011342A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant RNF13.

The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. The specific function of this gene has not yet been determined. Alternatively spliced transcript variants that encode the same protein have been reported. A pseudogene, which is also located on chromosome 3, has been defined for this gene. [provided by RefSeq

Sequence: LPARFGYRLPAEGLKGFLINSKPENACEPIVPPPVKDNSSGTFIVLIRRLDCNFDIKVLNAQRAGYKAAIVHNVDSDDLISMGSNDIEVLKKIDIPSVFI

Specifications

Antigen RNF13
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant RNF13.
Formulation 50% glycerol
Gene RNF13
Gene Accession No. NM_007282
Gene Alias FLJ93817/MGC13689/RZF
Gene Symbols RNF13
Host Species Mouse
Immunogen RNF13 (NP_009213, 51 a.a. ∽ 150 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11342
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.