Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNH1, Mouse, Clone: AT1H23, Abnova™

Catalog No. 89112915 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-915 Supplier Abnova Corporation Supplier No. MAB6508
Add to Cart
Add to Cart

Mouse monoclonal antibody raised aganist partial recombinant RNH1.

Specifications

Antigen RNH1
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT1H23
Conjugate Unconjugated
Description Mouse monoclonal antibody raised aganist partial recombinant RNH1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene RNH1
Gene Alias MGC18200/MGC4569/MGC54054/RAI/RNH
Gene Symbols RNH1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 7-461 of human RNH1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6050
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.