Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotavirus, Mouse, Clone: B197M, Abnova™

Catalog No. 89135852 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-852 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-852 Supplier Abnova Corporation Supplier No. MAB4889
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against rotavirus.

Specifications

Antigen Rotavirus
Applications ELISA, Immunofluorescence, Sandwich ELISA
Classification Monoclonal
Clone B197M
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against rotavirus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified Rhesus Rotavirus (RRV).
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with intact virus of strains RRV, WA, SA-11 and bovine. Immunostains infected cells. Does not react with Influenza A, Influenza B, RSV, Parainfluenza 1, 2 & 3, Adenovirus, M pneumonia, H. pylori and Mammalian cells.
Target Species Bacteria, Virus
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.