Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPA1, Mouse, Clone: 2H10, 70C or alphaSSB70C, Abnova™

Catalog No. 89111402 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-402 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-402 Supplier Abnova Corporation Supplier No. MAB3410

Mouse monoclonal antibody raised against RPA1.

Specifications

Antigen RPA1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2H10, 70C or alphaSSB70C
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against RPA1.
Dilution Immunoprecipitation (2 ug/mg) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide)
Gene RPA1
Gene Alias HSSB/REPA1/RF-A/RP-A/RPA70
Gene Symbols RPA1
Host Species Mouse
Immunogen Purified human RPA protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6117
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.