Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPE, Mouse, Clone: 401.8B11.3D9, Abnova™

Catalog No. 89110989 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-989 Supplier Abnova Corporation Supplier No. MAB2397
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native RPE65.

Specifications

Antigen RPE
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 401.8B11.3D9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native RPE65.
Dilution Immunohistochemistry (Frozen sections) (1:250) Western Blot (1:5000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene RPE65
Gene Symbols RPE65
Host Species Mouse
Immunogen Native purified from bovine RPE microsomal membrane proteins.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282043
Test Specificity This antibody is specific to RPE65.
Target Species Bovine, Human, Mouse, Pig
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.