Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL15, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105351 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-351 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-351 Supplier Abnova Corporation Supplier No. H00006138A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant RPL15.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L15E family of ribosomal proteins. It is located in the cytoplasm. This gene shares sequence similarity with the yeast ribosomal protein YL10 gene. Although this gene has been referred to as RPL10, its official symbol is RPL15. This gene has been shown to be overexpressed in some esophageal tumors compared to normal matched tissues. Transcript variants utilizing alternative polyA signals exist. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq

Sequence: RAGRHCGALRVLNSYWVGEDSTYKFFEVILIDPFHKAIRRNPDTQWITKPVHKHREMRGLTSAGRKSRGLGKGHKFHHTIGGSRRAAWRRRNTLQLHRY

Specifications

Antigen RPL15
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant RPL15.
Formulation 50% glycerol
Gene RPL15
Gene Accession No. NM_002948
Gene Alias EC45/FLJ26304/MGC88603/RPL10/RPLY10/RPYL10
Gene Symbols RPL15
Host Species Mouse
Immunogen RPL15 (NP_002939, 105 a.a. ∽ 203 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6138
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.