Learn More
RPS17, Mouse, Clone: 2C7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RPS17.
Supplier: Abnova Corporation H00006218M01A
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S17E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq
Sequence: EEIAIIPSKKLRNKIAGYVTHLMKRIQRGPVRGISIKLQEEERERRDNYVPEVSALDQEIIEVDPDTKEMLKLLDFGSLSNLQVTQPTVGMNFKTPRGPVSpecifications
RPS17 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_001021 | |
RPS17 | |
RPS17 (NP_001012,36 a.a. ∽ 135 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
6218 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
2C7 | |
Mouse monoclonal antibody raised against a partial recombinant RPS17. | |
RPS17 | |
DBA4/MGC72007/RPS17L1/RPS17L2 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.