Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS27A, Mouse, Clone: Ubi-1, Abnova™

Catalog No. 89135176 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-176 Supplier Abnova Corporation Supplier No. MAB2414
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against RPS27A.

Specifications

Antigen RPS27A
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone Ubi-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against RPS27A.
Dilution Immunofluorescence (1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 5mM sodium azide
Host Species Mouse
Immunogen Purified ubiquitin conjugated with KLH with glutaraldehyde.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody specifically recognizes ubiquitinated cytoplasmic inclusion bodies.
Target Species Bovine, Chicken, Drosophila, Human, Mouse, Nematode, Plant
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.