Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

rRNA, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89135175 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-175 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-175 Supplier Abnova Corporation Supplier No. MAB2412
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native rRNA.

Specifications

Antigen rRNA
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native rRNA.
Dilution Immunoprecipitation (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 1M Tris, 1.5M NaCl, 100mM glycine (0.05% sodium azide)
Host Species Mouse
Immunogen Native purified 5.8S ribosomal RNA.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to ribosomal RNA. There is no cross-reactivity with other RNAs.
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.