Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RYK, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105383 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-383 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-383 Supplier Abnova Corporation Supplier No. H00006259A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant RYK.

The protein encoded by this gene is an atypical member of the family of growth factor receptor protein tyrosine kinases, differing from other members at a number of conserved residues in the activation and nucleotide binding domains. This gene product belongs to a subfamily whose members do not appear to be regulated by phosphorylation in the activation segment. It has been suggested that mediation of biological activity by recruitment of a signaling-competent auxiliary protein may occur through an as yet uncharacterized mechanism. Two alternative splice variants have been identified, encoding distinct isoforms. [provided by RefSeq

Sequence: ELQSASAGPSVSLYLSEDEVRRLIGLDAELYYVRNDLISHYALSFNLLVPSETNFLHFTWHAKSKVEYKLGFQVDNVLAMDMPQVNISVQGEVPRTLSVFRVELSCTGKV

Specifications

Antigen RYK
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant RYK.
Formulation 50% glycerol
Gene RYK
Gene Accession No. NM_002958
Gene Alias D3S3195/JTK5/JTK5A/RYK1
Gene Symbols RYK
Host Species Mouse
Immunogen RYK (NP_002949, 54 a.a. ∽ 163 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6259
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.