Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100 Mouse anti-Bovine, Human, Mouse, Rat, Clone: 4C4.9, Abnova™

Catalog No. 89135437 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-437 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-437 Supplier Abnova Corporation Supplier No. MAB3352
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native S100.

Specifications

Antigen S100
Applications Immunohistochemistry
Classification Monoclonal
Clone 4C4.9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native S100.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified S100 from bovine brain.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.