Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100, Mouse, Clone: S100 217, Abnova™

Catalog No. 89136146 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-146 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-146 Supplier Abnova Corporation Supplier No. MAB6197
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against S100.

Specifications

Antigen S100
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone S100 217
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against S100.
Dilution Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (1% BSA, 0.05% sodium azide)
Host Species Mouse
Immunogen Purified bovine S100.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.